accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

HOOOOH

CAS RN 501-30-4

IUPAC Name: 5-hydroxy-2-(hydroxymethyl)-4H-pyran-4-one
Synonyms: kojic acid acido kojico kojyl-APPA kojisaeure
Molecular Weight (g/mol): 142.11
Molecular Formula: C6H6O4
InChi Key: BEJNERDRQOWKJM-UHFFFAOYSA-N
SMILES: OCC1=CC(=O)C(O)=CO1
Quantity 
  • (1)
  • (1)
  • (2)
  • (3)
  • (3)
  • (1)
Percent Purity 
  • (3)
  • (7)
Physical Form 
  • (5)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (4)
  • (3)
  • (1)
  • (1)
  • (2)
  • (3)
  • (3)
  • (1)
  • (3)
  • (7)
  • (5)