accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 501-53-1

IUPAC Name: benzyl carbonochloridate
Synonyms: benzyl chloroformate
Molecular Weight (g/mol): 170.59
Molecular Formula: C8H7ClO2
InChi Key: HSDAJNMJOMSNEV-UHFFFAOYSA-N
SMILES: ClC(=O)OCC1=CC=CC=C1
Quantity 
  • (1)
  • (3)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
Molecular Weight (g/mol) 
  • (1)
  • (3)
  • (7)
Percent Purity 
  • (3)
  • (3)
  • (4)
Boiling Point 
  • (1)
  • (3)
  • (4)
Physical Form 
  • (5)
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (3)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (3)
  • (7)
  • (3)
  • (3)
  • (4)
  • (1)
  • (3)
  • (4)
  • (5)
  • (3)