accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

HOOOH(E)

CAS RN 501-98-4

IUPAC Name: (2E)-3-(4-hydroxyphenyl)prop-2-enoic acid
Synonyms: para-coumaric acid p-cumaric acid trans-p-coumarinic acid 4'-hydroxycinnamate 4-coumaric acid, (z)-isomer naringeninic acid 4-coumaric acid, (E)-isomer (E)-isomer 4-coumaric acid (z)-isomer 4-coumaric acid 4-coumaric acid, (Z)-
Molecular Weight (g/mol): 164.16
Molecular Formula: C9H8O3
InChi Key: NGSWKAQJJWESNS-ZZXKWVIFSA-N
SMILES: OC(=O)\C=C\C1=CC=C(O)C=C1
Percent Purity 
  • (3)
  • (4)
Quantity 
  • (1)
  • (1)
  • (2)
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results