accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 502182-56-1

IUPAC Name: methyl 2-(morpholine-4-sulfonyl)benzoate
Synonyms: methyl 2-(morpholine-4-sulfonyl)benzoate
Molecular Weight (g/mol): 285.31
Molecular Formula: C12H15NO5S
InChi Key: FJLVVZZMDOHEEM-UHFFFAOYSA-N
SMILES: COC(=O)C1=CC=CC=C1S(=O)(=O)N1CCOCC1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results