accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 5029-67-4

IN

CAS RN 5029-67-4

IUPAC Name: 2-iodopyridine
Synonyms: 2-iodopyridine
Molecular Weight (g/mol): 205
Molecular Formula: C5H4IN
InChi Key: CCZWSTFVHJPCEM-UHFFFAOYSA-N
SMILES: IC1=CC=CC=N1
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (1)
  • (1)
  • (2)
Boiling Point 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
  • (1)
  • (2)
  • (2)
  • (2)