accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 502935-47-9

IUPAC Name: diphenyl-1,3-thiazole-5-carboxylic acid
Synonyms: diphenyl-1,3-thiazole-5-carboxylic acid
Molecular Weight (g/mol): 281.33
Molecular Formula: C16H11NO2S
InChi Key: KMOCHRNIGWCEJV-UHFFFAOYSA-N
SMILES: OC(=O)C1=C(N=C(S1)C1=CC=CC=C1)C1=CC=CC=C1
Molecular Weight (g/mol) 
  • (3)
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (3)
  • (3)
  • (3)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)