accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 503-87-7

IUPAC Name: 2-sulfanylideneimidazolidin-4-one
Synonyms: 4-imidazolidinone, 2-thioxo-
Molecular Weight (g/mol): 116.14
Molecular Formula: C3H4N2OS
InChi Key: UGWULZWUXSCWPX-UHFFFAOYSA-N
SMILES: O=C1CNC(=S)N1
Quantity 
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
Percent Purity 
  • (2)
  • (1)
Physical Form 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (3)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (1)