accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

ONOHNO

CAS RN 50332-66-6

IUPAC Name: 3-nitro-1,4-dihydroquinolin-4-one
Synonyms: 3-nitro-1H-quinolin-4-one
Molecular Weight (g/mol): 190.16
Molecular Formula: C9H6N2O3
InChi Key: ZWISCKSGNCMAQO-UHFFFAOYSA-N
SMILES: [O-][N+](=O)C1=CNC2=CC=CC=C2C1=O
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results