CAS RN 5034-06-0
CAS RN 5034-06-0
IUPAC Name:
trimethyl(oxo)-λ⁶-sulfanylium chloride
Synonyms:
trimethyl(oxo)-λ⁶-sulfanylium chloride
Molecular Weight (g/mol):
128.61
Molecular Formula:
C3H9ClOS
InChi Key:
KQYWHJICYXXDSQ-UHFFFAOYSA-M
SMILES:
[Cl-].C[S+](C)(C)=O
1
–
1
of
1
results
1
Trimethylsulfoxonium chloride, 97%
CAS: 5034-06-0 Molecular Formula: C3H9ClOS Molecular Weight (g/mol): 128.62 MDL Number: MFCD00011898 InChI Key: KQYWHJICYXXDSQ-UHFFFAOYSA-M Synonym: trimethylsulfoxonium chloride,trimethyloxosulfonium chloride,trimethylsulfonium chloride oxide,trimethyloxosulphonium chloride,sulfonium, trimethyl-, chloride, s-oxide,sulfoxonium, trimethyl-, chloride,sulfoxonium, trimethyl-, chloride 8ci,9ci,trimethyl oxo-??-sulfanylium chloride,c3h9os.cl,sulfoxonium, chloride PubChem CID: 197819 SMILES: C[S+](=O)(C)C.[Cl-]