accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

H2NNN

CAS RN 5036-48-6

IUPAC Name: 3-(1H-imidazol-1-yl)propan-1-amine
Synonyms: 3-(imidazol-1-yl)propan-1-amine
Molecular Weight (g/mol): 125.18
Molecular Formula: C6H11N3
InChi Key: KDHWOCLBMVSZPG-UHFFFAOYSA-N
SMILES: NCCCN1C=CN=C1
Boiling Point 
  • (2)
  • (2)
Quantity 
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results