accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 50397-74-5

NH2BrN

CAS RN 50397-74-5

IUPAC Name: 4-amino-3-bromobenzonitrile
Synonyms: 4-amino-3-bromobenzonitrile
Molecular Weight (g/mol): 197.04
Molecular Formula: C7H5BrN2
InChi Key: POESQIHWIIWNJL-UHFFFAOYSA-N
SMILES: NC1=CC=C(C=C1Br)C#N
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results