accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

OONHSHN

CAS RN 504-17-6

IUPAC Name: 2-sulfanylidene-1,3-diazinane-4,6-dione
Synonyms: thiobarbituric acid 2-thiobarbituric acid bathyran usaf EK-660 2-mercaptobarbituric acid austranal
Molecular Weight (g/mol): 144.15
Molecular Formula: C4H4N2O2S
InChi Key: RVBUGGBMJDPOST-UHFFFAOYSA-N
SMILES: O=C1CC(=O)NC(=S)N1
Percent Purity 
  • (3)
  • (3)
Quantity 
  • (2)
  • (1)
  • (1)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results