accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

H3CNCH3OH

CAS RN 50438-75-0

IUPAC Name: 2-[4-(dimethylamino)phenyl]ethan-1-ol
Synonyms: 2-[4-(dimethylamino)phenyl]ethanol
Molecular Weight (g/mol): 165.24
Molecular Formula: C10H15NO
InChi Key: CDTPAAZQBPSVGS-UHFFFAOYSA-N
SMILES: CN(C)C1=CC=C(CCO)C=C1
Boiling Point 
  • (2)
Molecular Weight (g/mol) 
  • (1)
  • (1)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)