accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

H3CSClOHO

CAS RN 50451-84-8

IUPAC Name: 5-chloro-3-methyl-1-benzothiophene-2-carboxylic acid
Synonyms: 5-chloro-3-methyl-1-benzothiophene-2-carboxylic acid
Molecular Weight (g/mol): 226.67
Molecular Formula: C10H7ClO2S
InChi Key: KEYCXZNZFABITO-UHFFFAOYSA-N
SMILES: CC1=C(SC2=CC=C(Cl)C=C12)C(O)=O
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results