accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

H3CSOH

CAS RN 505-10-2

IUPAC Name: 3-(methylsulfanyl)propan-1-ol
Synonyms: methionol
Molecular Weight (g/mol): 106.18
Molecular Formula: C4H10OS
InChi Key: CZUGFKJYCPYHHV-UHFFFAOYSA-N
SMILES: CSCCCO
Molecular Weight (g/mol) 
  • (3)
  • (3)
Quantity 
  • (1)
  • (2)
  • (1)
  • (1)
Boiling Point 
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (3)
  • (2)
  • (3)
  • (3)
  • (1)
  • (2)
  • (1)
  • (1)
  • (3)
  • (1)