accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 505083-04-5

CH3OOFBOHHO

CAS RN 505083-04-5

IUPAC Name: [3-fluoro-4-(methoxycarbonyl)phenyl]boronic acid
Synonyms: 3-fluoro-4-(methoxycarbonyl)phenylboronic acid
Molecular Weight (g/mol): 197.96
Molecular Formula: C8H8BFO4
InChi Key: YZYGXFXSMDUXJT-UHFFFAOYSA-N
SMILES: COC(=O)C1=CC=C(C=C1F)B(O)O
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results