accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 50525-27-4

IUPAC Name: λ²-ruthenium(2+) tris(1,10-phenanthroline) hexahydrate dichloride
Synonyms: λ²-ruthenium(2+) tris(phen) hexahydrate dichloride
Molecular Weight (g/mol): 820.69
Molecular Formula: C36H36Cl2N6O6Ru
InChi Key: UUSPGQXHSZVVNL-UHFFFAOYSA-L
SMILES: O.O.O.O.O.O.[Cl-].[Cl-].[Ru++].C1=CN=C2C(C=CC3=CC=CN=C23)=C1.C1=CN=C2C(C=CC3=CC=CN=C23)=C1.C1=CN=C2C(C=CC3=CC=CN=C23)=C1
Physical Form 
  • (2)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (1)
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)