accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 50551-58-1

IUPAC Name: ethyl 7-methoxy-1-benzofuran-2-carboxylate
Synonyms: ethyl 7-methoxy-1-benzofuran-2-carboxylate
Molecular Weight (g/mol): 220.22
Molecular Formula: C12H12O4
InChi Key: SXCMZYVXJUJIDJ-UHFFFAOYSA-N
SMILES: CCOC(=O)C1=CC2=CC=CC(OC)=C2O1
Quantity 
  • (2)
  • (1)
  • (1)
Physical Form 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results