accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

H3COHO(E)(E)(E)(E)

CAS RN 506-32-1

IUPAC Name: (5E,8E,11E,14E)-icosa-5,8,11,14-tetraenoic acid
Synonyms: arachidonic acid
Molecular Weight (g/mol): 304.47
Molecular Formula: C20H32O2
InChi Key: YZXBAPSDXZZRGB-CGRWFSSPSA-N
SMILES: CCCCC\C=C\C\C=C\C\C=C\C\C=C\CCCC(O)=O
Molecular Weight (g/mol) 
  • (8)
  • (1)
Quantity 
  • (2)
  • (1)
  • (1)
  • (2)
  • (2)
  • (1)
Percent Purity 
  • (1)
  • (5)
Boiling Point 
  • (1)
Grade 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (5)
  • (1)
  • (3)
  • (5)
  • (8)
  • (1)
  • (2)
  • (1)
  • (1)
  • (2)
  • (2)
  • (1)
  • (1)
  • (5)
  • (1)
  • (1)
  • (1)