CAS RN 506-44-5
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
(9Z,12Z,15Z)-Octadeca-9,12,15-trien-1-ol (>90%), TRC
CAS: 506-44-5 Molecular Formula: C18 H32 O Molecular Weight (g/mol): 264.45 Synonym: Linolenyl Alcohol,(9Z,12Z,15Z)-9,12,15-Octadecatrien-1-ol,(9Z,12Z,15Z)-α-Linolen-1-ol,(Z,Z,Z)-9,12,15-Octadecatrien-1-ol,(Z,Z,Z)-9,12,15-Octadecatriene-1-ol,9,12,15-Octadecatrienol,9-cis,12-cis-15-cis-Octadecatrienol,Linolenic Alcohol,Linolenyl Alcohol,all-cis-9,12,15-Octadecatrien-1-ol,cis,cis,cis-9,12,15-Octadecatrienol,cis-1-Hydroxy-9,12,15-Octadecatriene,α-Linolenol,α-Linolenyl Alcohol; IUPAC Name: (9Z,12Z,15Z)-octadeca-9,12,15-trien-1-ol SMILES: CC\C=C/C\C=C/C\C=C/CCCCCCCCO