accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

HClH3CNHCH3

CAS RN 506-59-2

IUPAC Name: hydrogen dimethylamine chloride
Synonyms: hydrogen dimethylamine chloride
Molecular Weight (g/mol): 81.54
Molecular Formula: C2H8ClN
InChi Key: IQDGSYLLQPDQDV-UHFFFAOYSA-N
SMILES: [H+].[Cl-].CNC
Molecular Weight (g/mol) 
  • (5)
  • (3)
  • (3)
Percent Purity 
  • (3)
  • (3)
  • (4)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
Physical Form 
  • (4)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (4)
  • (1)
  • (4)
  • (3)
  • (3)
  • (5)
  • (3)
  • (3)
  • (3)
  • (3)
  • (4)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
  • (4)