accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

ONHCl

CAS RN 50606-58-1

IUPAC Name: hydrogen 1-benzylpiperidin-3-one chloride
Synonyms: hydrogen 1-benzylpiperidin-3-one chloride
Molecular Weight (g/mol): 225.72
Molecular Formula: C12H16ClNO
InChi Key: OVWSFXNSJDMRPV-UHFFFAOYSA-N
SMILES: [H+].[Cl-].O=C1CCCN(CC2=CC=CC=C2)C1
Quantity 
  • (2)
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (3)
  • (2)
  • (2)
  • (2)
  • (3)