accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 50654-94-9

H3CH3CCH3OONH(R)NHNOO

CAS RN 50654-94-9

IUPAC Name: (2R)-2-{[(tert-butoxy)carbonyl]amino}-3-(1H-imidazol-5-yl)propanoate
Synonyms: (2R)-2-[(tert-butoxycarbonyl)amino]-3-(3H-imidazol-4-yl)propanoate
Molecular Weight (g/mol): 254.27
Molecular Formula: C11H16N3O4
InChi Key: AYMLQYFMYHISQO-MRVPVSSYSA-M
SMILES: CC(C)(C)OC(=O)N[C@H](CC1=CN=CN1)C([O-])=O
Molecular Weight (g/mol) 
  • (3)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (3)
  • (3)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)