accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CH3S

CAS RN 5069-26-1

IUPAC Name: 2-methyl-5-phenylthiophene
Synonyms: 2-methyl-5-phenylthiophene
Molecular Weight (g/mol): 174.26
Molecular Formula: C11H10S
InChi Key: NEKXRPVOSZHILM-UHFFFAOYSA-N
SMILES: CC1=CC=C(S1)C1=CC=CC=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results