accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

BrSBrO

CAS RN 507-16-4

IUPAC Name: sulfurooyl dibromide
Synonyms: thionyl bromide
Molecular Weight (g/mol): 207.87
Molecular Formula: Br2OS
InChi Key: HFRXJVQOXRXOPP-UHFFFAOYSA-N
SMILES: BrS(Br)=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results