accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 5070-13-3

IUPAC Name: bis(4-nitrophenyl) carbonate
Synonyms: bis(4-nitrophenyl) carbonate
Molecular Weight (g/mol): 304.21
Molecular Formula: C13H8N2O7
InChi Key: ACBQROXDOHKANW-UHFFFAOYSA-N
SMILES: [O-][N+](=O)C1=CC=C(OC(=O)OC2=CC=C(C=C2)[N+]([O-])=O)C=C1
Molecular Weight (g/mol) 
  • (6)
  • (2)
Quantity 
  • (1)
  • (1)
  • (3)
  • (2)
  • (1)
Percent Purity 
  • (3)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results