accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

H3CCH3OON

CAS RN 50743-32-3

IUPAC Name: 4-oxo-6-(propan-2-yl)-4H-chromene-3-carbonitrile
Synonyms: 6-isopropyl-4-oxochromene-3-carbonitrile
Molecular Weight (g/mol): 213.24
Molecular Formula: C13H11NO2
InChi Key: IMVAJLIIWCJMJP-UHFFFAOYSA-N
SMILES: CC(C)C1=CC=C2OC=C(C#N)C(=O)C2=C1
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (5)
  • (5)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)