accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

OFFFONHH2NOFFF

CAS RN 50778-75-1

IUPAC Name: 2,5-bis(2,2,2-trifluoroethoxy)benzohydrazide
Synonyms: 2,5-bis(2,2,2-trifluoroethoxy)benzohydrazide
Molecular Weight (g/mol): 332.2
Molecular Formula: C11H10F6N2O3
InChi Key: WJYCMPVGQTYGSQ-UHFFFAOYSA-N
SMILES: NNC(=O)C1=C(OCC(F)(F)F)C=CC(OCC(F)(F)F)=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results