accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 50868-72-9

IUPAC Name: 5-methoxy-2-methylaniline
Synonyms: 2-methyl-5-ethoxyaniline
Molecular Weight (g/mol): 137.18
Molecular Formula: C8H11NO
InChi Key: RPJXLEZOFUNGNZ-UHFFFAOYSA-N
SMILES: COC1=CC=C(C)C(N)=C1
Molecular Weight (g/mol) 
  • (3)
  • (4)
Quantity 
  • (3)
  • (1)
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results