accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 50890-83-0

IUPAC Name: 1-methyl-1H-indazole-3-carboxylic acid
Synonyms: 1-methylindazole-3-carboxylic acid
Molecular Weight (g/mol): 176.18
Molecular Formula: C9H8N2O2
InChi Key: OVVDFORZEGKEJM-UHFFFAOYSA-N
SMILES: CN1N=C(C(O)=O)C2=CC=CC=C12
Molecular Weight (g/mol) 
  • (4)
  • (2)
Percent Purity 
  • (3)
  • (2)
Quantity 
  • (2)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results