accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 50893-53-3

CH3ClOClO

CAS RN 50893-53-3

IUPAC Name: 1-chloroethyl carbonochloridate
Synonyms: chloroethyl chloroformate
Molecular Weight (g/mol): 142.96
Molecular Formula: C3H4Cl2O2
InChi Key: QOPVNWQGBQYBBP-UHFFFAOYNA-N
SMILES: CC(Cl)OC(Cl)=O
Molecular Weight (g/mol) 
  • (4)
  • (1)
Quantity 
  • (1)
  • (1)
  • (2)
  • (1)
Boiling Point 
  • (2)
  • (2)
Percent Purity 
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (1)
  • (4)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)