accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 50926-11-9

OSnIn

CAS RN 50926-11-9

IUPAC Name: stannanone indium
Synonyms: tin(II) oxide indium
Molecular Weight (g/mol): 249.53
Molecular Formula: InOSn
InChi Key: AMGQUBHHOARCQH-UHFFFAOYSA-N
SMILES: [In].O=[Sn]
Physical Form 
  • (3)
  • (5)
Quantity 
  • (3)
  • (3)
  • (2)
Percent Purity 
  • (2)
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (8)
  • (3)
  • (5)
  • (3)
  • (3)
  • (2)
  • (2)
  • (3)