accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 5098-14-6

IUPAC Name: 2-aminopropanedinitrile; 4-methylbenzene-1-sulfonic acid
Synonyms: 2-aminopropanedinitrile; para-toluene sulfonate
Molecular Weight (g/mol): 253.28
Molecular Formula: C10H11N3O3S
InChi Key: MEUWQVWJLLBVQI-UHFFFAOYSA-N
SMILES: NC(C#N)C#N.CC1=CC=C(C=C1)S(O)(=O)=O
Molecular Weight (g/mol) 
  • (2)
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results