accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

NNBr

CAS RN 50998-17-9

IUPAC Name: 6-bromoquinoxaline
Synonyms: 6-bromoquinoxaline
Molecular Weight (g/mol): 209.05
Molecular Formula: C8H5BrN2
InChi Key: NOYFLUFQGFNMRB-UHFFFAOYSA-N
SMILES: BrC1=CC=C2N=CC=NC2=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results