accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CH3NNHOCl(E/Z)

CAS RN 51-15-0

IUPAC Name: {[(2E)-1-methyl-1,2-dihydropyridin-2-ylidene]methyl}(oxo)azanium chloride
Synonyms: pralidoxime chloride
Molecular Weight (g/mol): 172.61
Molecular Formula: C7H9ClN2O
InChi Key: HIGSLXSBYYMVKI-UHDJGPCESA-N
SMILES: [Cl-].CN1C=CC=C\C1=C/[NH+]=O
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results