accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 51-36-5

IUPAC Name: 3,5-dichlorobenzoic acid
Synonyms: 3,5-dichlorobenzoic acid
Molecular Weight (g/mol): 191.01
Molecular Formula: C7H4Cl2O2
InChi Key: CXKCZFDUOYMOOP-UHFFFAOYSA-N
SMILES: OC(=O)C1=CC(Cl)=CC(Cl)=C1
Quantity 
  • (1)
  • (1)
  • (1)
Concentration 
  • (1)
Solvent or Matrix 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results