accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

HOOClCl

CAS RN 51-44-5

IUPAC Name: 3,4-dichlorobenzoic acid
Synonyms: 3,4-dichlorobenzoic acid
Molecular Weight (g/mol): 191.01
Molecular Formula: C7H4Cl2O2
InChi Key: VPHHJAOJUJHJKD-UHFFFAOYSA-N
SMILES: OC(=O)C1=CC=C(Cl)C(Cl)=C1
Percent Purity 
  • (1)
  • (1)
  • (3)
Quantity 
  • (1)
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results