accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 51-60-5

H3COSOOOCH3NH3COONH3CH3CCH3

CAS RN 51-60-5

IUPAC Name: 3-[(dimethylcarbamoyl)oxy]-N,N,N-trimethylanilinium methyl sulfate
Synonyms: neostigmine methylsulfate
Molecular Weight (g/mol): 334.39
Molecular Formula: C13H22N2O6S
InChi Key: OSZNNLWOYWAHSS-UHFFFAOYSA-M
SMILES: COS([O-])(=O)=O.CN(C)C(=O)OC1=CC=CC(=C1)[N+](C)(C)C
Quantity 
  • (1)
  • (1)
  • (1)
Certifications/Compliance 
  • (1)
  • (1)
Industry Type 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results