accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

NH2OH

CAS RN 51-67-2

IUPAC Name: 4-(2-aminoethyl)phenol
Synonyms: tyramine tyrosamine P-hydroxyphenethylamine 4-hydroxy-b-phenylethylamine tyramine base tenosin-wirkstoff p-tyramine P-hydroxy-β-phenethylamine P-hydroxy-b-phenylethylamine 4-hydroxy-β-phenylethylamine
Molecular Weight (g/mol): 137.18
Molecular Formula: C8H11NO
InChi Key: DZGWFCGJZKJUFP-UHFFFAOYSA-N
SMILES: NCCC1=CC=C(O)C=C1
Molecular Weight (g/mol) 
  • (9)
  • (3)
Percent Purity 
  • (3)
  • (3)
  • (2)
Quantity 
  • (1)
  • (3)
  • (3)
  • (1)
Boiling Point 
  • (3)
  • (2)
Physical Form 
  • (2)
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (3)
  • (3)
  • (9)
  • (3)
  • (3)
  • (3)
  • (2)
  • (1)
  • (3)
  • (3)
  • (1)
  • (3)
  • (2)
  • (2)
  • (3)
  • (1)