accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 5100-34-5

IUPAC Name: ethyl 3-isocyanatopropanoate
Synonyms: ethyl 3-isocyanatopropanoate
Molecular Weight (g/mol): 143.14
Molecular Formula: C6H9NO3
InChi Key: XBSGYVHOINMTIM-UHFFFAOYSA-N
SMILES: CCOC(=O)CCN=C=O
Boiling Point 
  • (1)
  • (2)
Quantity 
  • (2)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results