accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

HOBOHO

CAS RN 51067-38-0

IUPAC Name: (4-phenoxyphenyl)boronic acid
Synonyms: 4-phenoxyphenylboronic acid
Molecular Weight (g/mol): 214.03
Molecular Formula: C12H11BO3
InChi Key: KFXUHRXGLWUOJT-UHFFFAOYSA-N
SMILES: OB(O)C1=CC=C(OC2=CC=CC=C2)C=C1
Molecular Weight (g/mol) 
  • (6)
  • (5)
Percent Purity 
  • (3)
  • (8)
Quantity 
  • (3)
  • (1)
  • (1)
  • (2)
  • (3)
  • (1)
Physical Form 
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (6)
  • (3)
  • (2)
  • (6)
  • (6)
  • (5)
  • (3)
  • (8)
  • (3)
  • (1)
  • (1)
  • (2)
  • (3)
  • (1)
  • (3)