accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

H3CONOOOOH

CAS RN 51073-04-2

IUPAC Name: 2-(2-methoxy-5-nitrophenyl)acetic acid
Synonyms: (2-methoxy-5-nitrophenyl)acetic acid
Molecular Weight (g/mol): 211.17
Molecular Formula: C9H9NO5
InChi Key: TZQZNDOEOCOJFS-UHFFFAOYSA-N
SMILES: COC1=C(CC(O)=O)C=C(C=C1)[N+]([O-])=O
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results