accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 51135-73-0

IUPAC Name: ethyl 5-methyl-1,2-oxazole-4-carboxylate
Synonyms: ethyl 5-methyl-1,2-oxazole-4-carboxylate
Molecular Weight (g/mol): 155.15
Molecular Formula: C7H9NO3
InChi Key: KOMSQTMQKWSQDW-UHFFFAOYSA-N
SMILES: CCOC(=O)C1=C(C)ON=C1
Boiling Point 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results