accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 51146-56-6

IUPAC Name: (2S)-2-[4-(2-methylpropyl)phenyl]propanoic acid
Synonyms: dexibuprofen dexibuprofenum d-ibuproten dexibuprofeno dexibuprofene
Molecular Weight (g/mol): 206.29
Molecular Formula: C13H18O2
InChi Key: HEFNNWSXXWATRW-JTQLQIEISA-N
SMILES: CC(C)CC1=CC=C(C=C1)[C@H](C)C(O)=O
Percent Purity 
  • (3)
  • (2)
Quantity 
  • (2)
  • (1)
  • (1)
  • (1)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (3)
  • (3)
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)
  • (2)