accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 51169-17-6

IUPAC Name: 5-(4-methylphenyl)-5-phenylimidazolidine-2,4-dione
Synonyms: 5-(4-methylphenyl)-5-phenylimidazolidine-2,4-dione
Molecular Weight (g/mol): 266.3
Molecular Formula: C16H14N2O2
InChi Key: WPAPSGQWYNPWCZ-UHFFFAOYNA-N
SMILES: CC1=CC=C(C=C1)C1(NC(=O)NC1=O)C1=CC=CC=C1
Percent Purity 
  • (3)
  • (1)
Quantity 
  • (2)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results