accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CH2ONO

CAS RN 5117-12-4

IUPAC Name: 1-(morpholin-4-yl)prop-2-en-1-one
Synonyms: acryloyl morpholine
Molecular Weight (g/mol): 141.17
Molecular Formula: C7H11NO2
InChi Key: XLPJNCYCZORXHG-UHFFFAOYSA-N
SMILES: C=CC(=O)N1CCOCC1
Boiling Point 
  • (2)
  • (2)
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)