accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 51175-71-4

SSOOCl

CAS RN 51175-71-4

IUPAC Name: thiophene-3-sulfonyl chloride
Synonyms: thiophene-3-sulfonyl chloride
Molecular Weight (g/mol): 182.64
Molecular Formula: C4H3ClO2S2
InChi Key: YSPWSQNKRBSICH-UHFFFAOYSA-N
SMILES: ClS(=O)(=O)C1=CSC=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results