accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 512-42-5

IUPAC Name: sodium methyl sulfate
Synonyms: methyl bisulfate
Molecular Weight (g/mol): 134.08
Molecular Formula: CH3NaO4S
InChi Key: DZXBHDRHRFLQCJ-UHFFFAOYSA-M
SMILES: [Na+].COS([O-])(=O)=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results