accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 512-56-1

IUPAC Name: trimethyl phosphate
Synonyms: trimethyl phosphate trimethylphosphate trimethyl orthophosphate ccris 610 phosphate, trimethyl- trimethyl-phosphate TMPA TMPO trimethylfosfat
Molecular Weight (g/mol): 140.08
Molecular Formula: C3H9O4P
InChi Key: WVLBCYQITXONBZ-UHFFFAOYSA-N
SMILES: COP(=O)(OC)OC
Molecular Weight (g/mol) 
  • (3)
  • (6)
Percent Purity 
  • (2)
  • (3)
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
Boiling Point 
  • (2)
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results