accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 5121-00-6

IUPAC Name: 2-(naphthalen-1-yl)acetyl chloride
Synonyms: 2-(naphthalen-1-yl)acetyl chloride
Molecular Weight (g/mol): 204.65
Molecular Formula: C12H9ClO
InChi Key: DSVAZLXLRDXHKO-UHFFFAOYSA-N
SMILES: ClC(=O)CC1=CC=CC2=CC=CC=C12
Quantity 
  • (2)
  • (1)
  • (1)
  • (1)
Boiling Point 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (2)
  • (1)
  • (1)
  • (1)
  • (2)